Learn More
Description
Specifications
Specifications
| Antigen | CD14 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | M5E2 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Simple Western 50 ug/ml, Flow Cytometry < 2.0 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, CyTOF-ready, Block/Neutralize reported in scientific literature (PMID 27228163), Dual RNAscope ISH-IHC |
| Gene Alias | CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein |
| Gene Symbols | CD14 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
