Learn More
Description
Ensure accurate, reproducible results in ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunofluorescence
Specifications
Specifications
| Antigen | CD27 Ligand/TNFSF7/CD70 |
| Applications | Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), Immunofluorescence |
| Classification | Monoclonal |
| Clone | TNFS7/1026 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunoprecipitation 0.5 - 1 ug/500ug Protein Lysate, Immunohistochemistry-Frozen 0.5 - 1 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml |
| Formulation | 10mM PBS and 0.05% BSA with 0.05% Sodium Azide |
| Gene Alias | CD27-L, CD27LCD27 ligand, CD27LG, CD70, CD70 molecule, Ki-24 antigen, surface antigen CD70, TNFSF7CD70 antigen, tumor necrosis factor (ligand) superfamily, member 7, Tumor necrosis factor ligand superfamily member 7 |
| Gene Symbols | CD70 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
