Learn More
Epredia™ Lab Vision™ CD3 (Early T-Cell Marker) Rabbit Monoclonal Antibody,
Stain normal and neoplastic T cells in formalin-fixed, paraffin-embedded tissues with Epredia™ CD3 (Early T-Cell Marker), Rabbit Monoclonal Antibody.
Specifications
| Antigen | CD3 |
|---|---|
| Clone | SP7 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
|
RM9107S0
|
Epredia™
RM9107S0 |
Concentrated | 100 μL | N/A | N/A | N/A | |||
|
RM9107S1
|
Epredia™
RM9107S1 |
Concentrated | 500 μL | N/A | N/A | N/A | |||
|
RM9107S
|
Epredia™
RM9107S |
Concentrated | 1 mL | N/A | N/A | N/A | |||
|
RM9107R7
|
Epredia™
RM9107R7 |
Prediluted | 7 mL | N/A | N/A | N/A | |||
|
RM9107RQ
|
Epredia™
RM9107RQ |
Ready-to-Use | 12 mL | N/A | N/A | N/A | |||
Description
This antibody reacts with the intracytoplasmic portion of the CD3 antigen expressed by T cells. It stains human T cells in both the cortex and medulla of the thymus and in peripheral lymphoid tissues. This antibody is suitable for staining normal and neoplastic T cells in formalin-fixed, paraffin-embedded tissues.Host Species: Rabbit
Clone: SP7
Isotype: IgG
Species Reactivity: Human, Baboon, Monkey, Horse, Dog and Cat. Others not tested.
Epitope: 156-168aa
Immunogen: A synthetic 13-mer peptide corresponding to aa 156-168 of the ε-chain of human CD3 protein
Positive Control: Jurkat cells, Tonsil
Cellular Localization: Membrane
Clone SP7 is excellent for staining of formal/paraffin.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| CD3 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| A synthetic 13-mer peptide corresponding to aa 156-168 of the epsilon-chain of human CD3 protein |
| SP7 | |
| Monoclonal | |
| Rabbit | |
| Cancer and Tumor Biology | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.