Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34 (Clone QBEnd/10) Ab-1 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry procedures with Epredia™ CD34 (Endothelial Cell Marker) Ab-1, Mouse Monoclonal Antibody.

$518.00 - $1279.00

Specifications

Antigen CD34 (Endothelial Cell Marker) Ab-1
Clone QBEnd/10
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Host Species Mouse
View More Specs

Products 3
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
MS363P
View Documents
Epredia
MS363P
Concentrated 1 mL
Each for $1,279.00
Only null left
Add to Cart
 
MS363R7
View Documents
Epredia
MS363R7
Prediluted 7 mL
Each for $518.00
Only null left
Add to Cart
 
MS363R1
missing translation for 'viewDocuments'
Epredia
MS363R1
Prediluted 1 mL N/A N/A N/A
Description

Description

CD34, a single chain transmembrane glycoprotein, is selectively expressed on human lymphoid and myeloid hematopoietic progenitor cells. Staining for CD34 has been used to measure angiogenesis, which reportedly predicts tumor recurrence. Antibody to CD34 also reacts with vascular endothelial cells in normal tissues and in benign and malignant proliferations. This antibody is of value in the study of benign and malignant vascular tumors as well as characterization of leukemias.

Host Species: Mouse

Clone: QBEnd/10

Isotype: IgG1

Species Reactivity: Human. Does not react with dog and rat. Others not known

Immunogen: Detergent solubilized vesicular suspension prepared from a perfusate of human term placenta

Molecular Weight: 105-120 kDa

Positive Control: Tonsil

Cellular Localization: Membrane/cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

CD34 (Endothelial Cell Marker) Ab-1
Immunohistochemistry (Paraffin)
Mouse
Oncogenesis
Detergent solubilized vesicular suspension prepared from a perfusate of human term placenta
Primary
QBEnd/10
Monoclonal
IVD
Human
IgG1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.