Learn More
CD86 Mouse anti-Rat, FITC, Clone: 24F, BD
Mouse Monoclonal Antibody
$128.00 - $384.00
Specifications
Antigen | CD86 |
---|---|
Clone | 24F |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The 24F antibody reacts with CD86 (B7-2), a member of the Ig superfamily of transmembrane proteins. CD86, a ligand for CD28 and CD152 (CTLA-4), is one of the accessory molecules that plays an important role in T cell-B cell costimulatory interactions. CD86 is predominantly expressed on antigen-presenting cells. It can be upregulated on splenic B cells by in vitro polyclonal activation, such as LPS treatment. 24F mAb is reported to block the costimulatory function of rat CD86 and to immunoprecipitate CD86 from Lewis-S1 cell lysates.HTLV-1-transformed rat T-cell line Lewis-S1
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
CD86 | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Rat | |
IgG1 κ | |
Affinity Purified | |
B7-2 |
24F | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.