Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD99/MIC2 (Ewing' s Sarcoma Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™

Detect CD99 in human and rat samples with Epredia™ CD99/MIC2 (Ewing's Sarcoma Marker) Ab-1, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

Specifications

Antigen CD99/MIC2 (Ewings Sarcoma Marker) Ab-1
Clone HO36-1.1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Catalog Number Mfr. No. Form Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Concentration Quantity Price Quantity & Availability  
MS294P0
View Documents
Epredia
MS294P0
Concentrated 200μg/mL 100 μL N/A N/A N/A
MS294P1
View Documents
Epredia
MS294P1
Concentrated 200μg/mL 500 μL N/A N/A N/A
MS294P
View Documents
Epredia
MS294P
Concentrated 200μg/mL 1 mL N/A N/A N/A
MS294P1ABX
View Documents
Epredia
MS294P1ABX
Concentrated 1mg/mL 100 μL N/A N/A N/A
MS294PABX
View Documents
Epredia
MS294PABX
Concentrated 1mg/mL 200 μL N/A N/A N/A
MS294R1
View Documents
Epredia
MS294R1
Prediluted 1 mL N/A N/A N/A
Description

Description

CD99, or MIC2 gene product, or E2 antigen is expressed on the cell membrane of some lymphocytes, cortical thymocytes, and granulosa cells of the ovary. The antigen is also expressed by most pancreatic islet cells, Sertoli cells of the testis, and some endothelial cells. Mature granulocytes express very little or no CD99. MIC2 is strongly expressed on Ewing's sarcoma cells and primitive peripheral neuroectodermal tumors.

Host Species: Mouse

Clone: HO36-1.1

Isotype: IgM

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified E-rosette forming cells from human peripheral blood lymphocytes

Molecular Weight: 27-32kDa

Positive Control: Pancreas or Ewing's sarcoma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Specifications

Specifications

CD99/MIC2 (Ewings Sarcoma Marker) Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
IVD
Human, Rat
IgM
HO36-1.1
Monoclonal
Mouse
Oncogenesis
Purified E-rosette forming cells from human peripheral blood lymphocytes
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.