Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDCA7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP182224 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP182224 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP182224 Supplier Novus Biologicals Supplier No. NBP182224

Rabbit Polyclonal Antibody has been used in 2 publications

CDCA7 Polyclonal specifically detects CDCA7 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CDCA7
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias cell division cycle associated 7, cell division cycle-associated protein 7, c-Myc target JPO1, FLJ14722, FLJ14736, JPO1cell division cycle associated protein 7, MGC34109, Protein JPO1
Gene Symbols CDCA7
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:PKFRSDISEELANVFYEDSDNESFCGFSESEVQDVLDHCGFLQKPRPDVTNELAGIFHADSDDESFCGFSESEIQDGM
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 83879
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.