Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDw75 Ab-1 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Epredia™ CDw75 Ab-1, Mouse Monoclonal Antibody.

$191.00 - $682.00

Specifications

Antigen CDw75 Ab-1
Clone LN1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Catalog Number Mfr. No. Form Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Concentration Quantity Price Quantity & Availability  
MS130P0
View Documents
Epredia
MS130P0
Concentrated 200μg/mL 100 μL
Each for $191.00
Only null left
Add to Cart
 
MS130P1
View Documents
Epredia
MS130P1
Concentrated 200μg/mL 500 μL
Each for $515.00
Only null left
Add to Cart
 
MS130P1ABX
View Documents
Epredia
MS130P1ABX
Concentrated 1mg/mL 100 μL
Each for $552.00
Only null left
Add to Cart
 
MS130PABX
View Documents
Epredia
MS130PABX
Concentrated 1mg/mL 200 μL
Each for $682.00
Only null left
Add to Cart
 
MS130P
View Documents
Epredia
MS130P
Concentrated 200μg/mL 1 mL N/A N/A N/A
MS130R1
View Documents
Epredia
MS130R1
Prediluted 1 mL N/A N/A N/A
MS130R7
View Documents
Epredia
MS130R7
Prediluted 7 mL N/A N/A N/A
Description

Description

CDw75 is a neuraminidase-sensitive sialoprotein, present on cell membrane and cytoplasm of germinal center B-cells and derived lymphomas. LN1 reacts with RBC precursors of bone marrow, ductal and ciliated epithelial cells of kidney, breast, prostate, pancreas, lung, and with glioblastomas, astrocytomas, and Reed Sternberg cells in lymphocyte predominant Hodgkin's disease. LN1 is shown to be a reliable antibody for ascribing a B-cell phenotype in known lymphoid tissues.

Host Species: Mouse

Clone: LN1

Isotype: IgM/κ

Species Reactivity: Human. Does not react with rat. Others not known

Epitope: Not determined

Immunogen: Nuclei from pokeweed mitogen-stimulated peripheral blood lymphocytes

Molecular Weight: 53 and 87 kDa

Positive Control: Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry

Specifications

Specifications

CDw75 Ab-1
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
IVD
Human
IgM κ
LN1
Monoclonal
Mouse
B-Cell
Nuclei from pokeweed mitogen-stimulated peripheral blood lymphocytes
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.