Learn More
Sonoco™ ThermoSafe™ Certis™ Platform Series 56-Liter Shippers
Certis series offers robust universal temperature control for temperature sensitive goods. The Certis solutions are material neutral, incorporating leading edge technologies to provide the lowest total cost of ownership.
$679.00 - $1508.00
Catalog Number | Mfr. No. | Duration | Applications | Dimensions (L x W x H) Exterior | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Duration | Applications | Dimensions (L x W x H) Exterior | Price | Quantity & Availability | |||
03-531-121
![]() |
Sonoco™
C56C3U |
72 hr. | Control Room Temperature (CRT) | 829 x 699 x 559 mm (32.625 x 27.5 x 22 in.) |
Each for $1,237.00
|
|
|||
03-531-122
|
Sonoco™
C56F3U |
72 hr. | Frozen | 829 x 699 x 559 mm (32.625 x 27.5 x 22 in.) |
Each for $679.00
|
|
|||
03-531-120
![]() |
Sonoco™
C56R3U |
72 hr. | Refrigerated | 829 x 699 x 559 mm (32.625 x 27.5 x 22 in.) |
Each for $1,150.00
|
|
|||
03-531-118
![]() |
Sonoco™
C56C5U |
120 hr. | Control Room Temperature (CRT) | 768 x 638 x 511 mm (30.25 x 25.13 x 20.13 in.) |
Each for $1,508.00
|
|
|||
03-531-119
![]() |
Sonoco™
C56F5U |
120 hr. | Frozen | 768 x 638 x 511 mm (30.25 x 25.13 x 20.13 in.) |
Each for $928.00
|
|
|||
03-531-117
![]() |
Sonoco™
C56R5U |
120 hr. | Refrigerated | 768 x 638 x 511 mm (30.25 x 25.13 x 20.13 in.) |
Each for $1,370.00
|
|
|||
Description
- All shippers are prequalified systems saving assembly time and space and optimize dim weight and freight cost
- Simply change the outer shipper to switch between 3-Day and 5-Day
- Switch between Refrigerated and CRT solutions by swapping Amber and Yellow bricks while keeping the same confi guration for the specific payload
- Same pack-out configurations across 72 hour and 120 hour durations
- Mistake-proof pack-out with color coded PCMs for Refrigerated (Amber) and CRT (Yellow)
- Payload dimensions: 603 x 394 x 235mm (23.75 x 15.5 x 9.25 in.)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Certis | |
55.8 L |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.