Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cetuximab Antibody (2259C) [Biotin], Novus Biologicals™
SDP

Catalog No. NB426976 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB426976 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB426976 Supplier Novus Biologicals Supplier No. FAB9626B
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cetuximab Monoclonal antibody specifically detects Cetuximab in Broad Species Reactivity samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA

Specifications

Antigen Cetuximab
Applications Western Blot, Simple Western, Flow Cytometry, ELISA
Classification Monoclonal
Clone 2259C
Conjugate Biotin
Dilution Western Blot, Simple Western, Flow Cytometry, ELISA
Formulation PBS
Host Species Rabbit
Immunogen Full length Cetuximab,
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Broad Species Reactivity
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.