Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Cevimeline hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
Selective M1 receptor agonist. Induces atropine-sensitive contractions of isolated guinea pig ilea and trachea preparations (EC50 values are 3.5 and 3 μM respectively). Reverses AF64A-induced cognitive impairments in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 235.77 |
| Purity | >98% |
| Molecular Weight (g/mol) | 235.77 |
| Chemical Name or Material | Cevimeline hydrochloride |
Safety and Handling
| Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction