Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CGP 35348
GREENER_CHOICE

Catalog No. p-4783328
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective GABAB antagonist; brain penetrant

Selective, brain penetrant GABAB receptor antagonist (IC50 = 34 μM as measured in rat cortical membranes). Has higher affinity for postsynaptic versus presynaptic receptors.

Chemical Identifiers

CAS 123690-79-9
Molecular Formula C8H20NO4P

Specifications

Quantity 10 mg
Formula Weight 225.22
Purity >98%
Molecular Weight (g/mol) 225.23
Chemical Name or Material CGP 35348
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.