Learn More
Eisco™ Chert Mineral Specimen
SCI_ED
Catalog No. p-7203008
Click to view available options
Quantity:
1/Ea.
12/Pk.
Sometimes referred to as flint, Chert is a sedimentary rock rich in silica.
  • Correlates well to Next Generation Science Standards Evidence
  • Chert is perfect for teaching about the many forms of quartz, hardness, crystal habit, and fracture
Specifications
Product Type Mineral
Grade K to 16
Quantity 1/Ea.
Specimen Size 2 to 3 cm
Weight (Metric) 0.1 kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.