Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLP1/HEAB Antibody (SR1834), Novus Biologicals™
SDP

Catalog No. NB259419 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259419 100 μg
NB258858 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB259419 Supplier Novus Biologicals Supplier No. NBP322477100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CLP1/HEAB Monoclonal antibody specifically detects CLP1/HEAB in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry

Specifications

Antigen CLP1/HEAB
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone SR1834
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias CLP1, cleavage and polyadenylation factor I subunit, homolog (S. cerevisiae), EC 2.7.1.78, hClp1, HEAB, HEABATP/GTP-binding protein, homolog of yeast CFIA subunit Clp1p, Polynucleotide kinase Clp1, polyribonucleotide 5'-hydroxyl-kinase Clp1, Pre-mRNA cleavage complex II protein Clp1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CLP1/HEAB (Uniprot #: Q92989)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10978
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.