Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C3aR Antibody (3G7) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7106042 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25267425 0.025 mg
NBP25267402 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25267425 Supplier Novus Biologicals Supplier No. NBP2526740.025MG
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Complement Component C3aR Monoclonal specifically detects Complement Component C3aR in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence, CyTOF-ready.

Specifications

Antigen Complement Component C3a R
Applications Flow Cytometry, ELISA, CyTOF
Classification Monoclonal
Clone 3G7
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, CyTOF-ready
Formulation PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Gene Alias AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, HNFAG09
Gene Symbols C3AR1
Host Species Rabbit
Immunogen The 2e loop of the C3a receptor.
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 719
Test Specificity Binds specifically to the 2e loop of the human C3a receptor, residues 185-193.
Target Species Human
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.