Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Conglomerate Specimen
SCI_ED

Catalog No. p-7076835 Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm

Conglomerate Specimen has large clasts that indicate that pieces of rock underwent large amount of rolling, bouncing, and other weathering to become round.

  • Large clasts within rock are ideal for teaching students about rounding and sorting
  • For use in teaching students about sorting and rounding
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Well-rounded and poorly sorted, indicating it was deposited and formed in running water

Specifications

Product Type Sedimentary Rock, Conglomerate
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.