Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COPS4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | COPS4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15664620
![]() |
Novus Biologicals
NBP15664620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP156646
![]() |
Novus Biologicals
NBP156646 |
100 μL |
Each for $487.50
|
|
|||||
Description
COPS4 Polyclonal specifically detects COPS4 in Human samples. It is validated for Western Blot.Specifications
COPS4 | |
Polyclonal | |
Rabbit | |
DNA Repair, Neuronal Cell Markers, Neurotransmission | |
COP9 constitutive photomorphogenic homolog subunit 4 (Arabidopsis), COP9 signalosome complex subunit 4, CSN4Arabidopsis, homolog) subunit 4, MANOP1, MGC10899, MGC15160 | |
COPS4 | |
Primary | |
46 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q9BT78 | |
51138 | |
IgG | |
This product is specific to Subunit or Isoform: 4. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title