Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX-2 Antibody, Novus Biologicals™
SDP

Catalog No. p-5059076 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100689SS 0.1 mL
NB100689 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100689SS Supplier Novus Biologicals Supplier No. NB100689SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 29 publications

COX-2 Polyclonal antibody specifically detects COX-2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).

Specifications

Antigen COX-2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Simple Western 1:100, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen
Formulation PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide
Gene Accession No. P35355
Gene Alias COX-2, COX2cyclooxygenase 2b, cyclooxygenase-2, EC 1.14.99, EC 1.14.99.1, GRIPGHS, hCox-2, PGG/HS, PGH synthase 2, PGHS-2, PHS II, PHS-2, prostaglandin G/H synthase 2, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 2, Prostaglandin-endoperoxide synthase 2, prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase andcyclooxygenase)
Gene Symbols PTGS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to the C-terminus of rat COX2 (the exact amino acid sequence is considered to be proprietary information).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Cytokine Research, Hypoxia, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5743
Test Specificity This reacts with the 70 kDa protein.
Target Species Human, Rat
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.