Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX6A1 Antibody (OTI4G2), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB276235 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB276235 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB276235 Supplier Novus Biologicals Supplier No. NBP270427MFV450
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COX6A1 Monoclonal antibody specifically detects COX6A1 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen COX6A1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI4G2
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias COX VIa-L, COX6A, COX6AL, Cytochrome c oxidase polypeptide VIa-liver, cytochrome c oxidase subunit 6A1, mitochondrial, cytochrome C oxidase subunit VIa homolog, cytochrome c oxidase subunit VIa polypeptide 1, Cytochrome c oxidase subunit VIA-liver, MGC104500
Host Species Mouse
Immunogen Full length human recombinant protein of human COX6A1 (NP_004364) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1337
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.