Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COX7B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | COX7B |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15702820
![]() |
Novus Biologicals
NBP15702820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP157028
![]() |
Novus Biologicals
NBP157028 |
100 μL |
Each for $487.50
|
|
|||||
Description
COX7B Polyclonal specifically detects COX7B in Human samples. It is validated for Western Blot.Specifications
COX7B | |
Polyclonal | |
Rabbit | |
P24311 | |
1349 | |
Synthetic peptides corresponding to COX7B (cytochrome c oxidase subunit VIIb) The peptide sequence was selected from the N terminal of COX7B. Peptide sequence MFPLVKSALNRLQVRSIQQTMARQSHQKRTPDFHDKYGNAVLASGATFCI. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Cytochrome c oxidase polypeptide VIIb, cytochrome c oxidase subunit 7B, mitochondrial, cytochrome c oxidase subunit VIIb, cytochrome-c oxidase chain VIIb | |
COX7B | |
IgG | |
This product is specific to Subunit or Isoform: 7B, mitochondrial. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title