Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatine Kinase, Muscle/CKMM Mouse anti-Human, Clone: CKMM/3341, Novus Biologicals™
SDP

Catalog No. NB122594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB122594 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB122594 Supplier Novus Biologicals Supplier No. NBP30756020UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Creatine Kinase, Muscle/CKMM Monoclonal specifically detects Creatine Kinase, Muscle/CKMM in Human samples. It is validated for ELISA, Protein Array.

Specifications

Antigen Creatine Kinase, Muscle/CKMM
Applications ELISA, Peptide Array
Classification Monoclonal
Clone CKMM/3341
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution ELISA, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Alias CKMMcreatine kinase M-type, Creatine Kinase Isoenzyme, Creatine kinase M chain, creatine kinase, muscle, creatine kinase-M, EC 2.7.3, EC 2.7.3.2, M-CK
Host Species Mouse
Immunogen Recombinant full-length Creatine Kinase, Muscle/CKMM protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1158
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.