Learn More
Creatine Kinase, Muscle/CKMM Mouse anti-Human, Clone: CKMM/3341, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Creatine Kinase, Muscle/CKMM |
| Applications | ELISA, Peptide Array |
| Classification | Monoclonal |
| Clone | CKMM/3341 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | ELISA, Protein Array |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | CKMMcreatine kinase M-type, Creatine Kinase Isoenzyme, Creatine kinase M chain, creatine kinase, muscle, creatine kinase-M, EC 2.7.3, EC 2.7.3.2, M-CK |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.