Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR3 (CD183) Armenian Hamster anti-Mouse, Unconjugated, Clone: 173, MilliporeSigma™
Armenian Hamster Monoclonal Antibody
$189.29 - $477.80
Specifications
Antigen | CXCR3 (CD183) |
---|---|
Clone | 173 |
Applications | Flow Cytometry, Inhibition Assays, Radioimmune Assays (RIA) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF21812MI
|
MilliporeSigma
MABF218125UL |
25 μL |
Each for $189.29
|
|
|||||
MABF21811MI
|
MilliporeSigma
MABF2181100UL |
100 μL |
Each for $477.80
|
|
|||||
Description
CXCR3 (CD183) Monoclonal antibody specifically detects CXCR3 (CD183) in Mouse samples. It is validated for Flow Cytometry, Inhibition, Radioimmunoassay.Specifications
CXCR3 (CD183) | |
Flow Cytometry, Inhibition Assays, Radioimmune Assays (RIA) | |
Unconjugated | |
Armenian Hamster | |
Mouse | |
C-X-C chemokine receptor type 3;CXC-R3;Interferon-inducible protein 10 receptor;IP-10 receptor;CD183 | |
A multiple antigen peptide (MAPS8) corresponding to 37 amino acids from the N-terminal region of murine C-X-C chemokine receptor 3 (CXCR3). | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
41.02 kDa calculated. |
173 | |
Monoclonal | |
Purified | |
Inflammation and Immunology | |
NP_034040 | |
Cxcr3, Cmkar3 | |
IgG | |
Protein G Purified | |
Clone 173 is an Armenian Hamster monoclonal antibody that specifically detects CXCR3 on cells from wild-type, but not from CXCR3-/- mice. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title