Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cyclin A Ab-6, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Specifications

Antigen Cyclin A Ab-6
Clone 6E6
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
MS1061S0
View Documents
Epredia™
MS1061S0
Concentrated 100 μL N/A N/A N/A
MS1061S1
View Documents
Epredia™
MS1061S1
Concentrated 500 μL N/A N/A N/A
MS1061S
View Documents
Epredia™
MS1061S
Concentrated 1 mL N/A N/A N/A
MS1061R7
View Documents
Epredia™
MS1061R7
Prediluted 7 mL N/A N/A N/A
Description

Description

Cyclins are regulatory subunits of the cyclin-dependent kinases (cdk's). They are expressed temporally to control transition at different specific phases of the cell cycle. In mammalian somatic cells, cyclin A is required for S-phase and passage through G2-phase.

Host Species: Mouse

Clone: 6E6

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: N-terminal

Immunogen: Recombinant N-terminal fragment of human cyclin A protein

Molecular Weight: 60 kDa

Positive Control: HeLa cells, Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin-fixed only)
  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cyclin A Ab-6
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
Human
IgG1 κ
6E6
Monoclonal
Mouse
Cell Cycle
Recombinant N-terminal fragment of human cyclin A protein
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.