Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP3A7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | CYP3A7 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP16238720
![]() |
Novus Biologicals
NBP16238720UL |
20 μL |
Each for $206.00
|
|
|||||
NBP162387
![]() |
Novus Biologicals
NBP162387 |
100 μL |
Each for $487.50
|
|
|||||
Description
CYP3A7 Polyclonal specifically detects CYP3A7 in Human samples. It is validated for Western Blot.Specifications
CYP3A7 | |
Polyclonal | |
Purified | |
RUO | |
P24462 | |
1551 | |
Synthetic peptides corresponding to CYP3A7(cytochrome P450, family 3, subfamily A, polypeptide 7) The peptide sequence was selected from the middle region of CYP3A7. Peptide sequence KSVKQIKEGRLKETQKHRVDFLQLMIDSQNSKDSETHKALSDLELMAQSI. | |
Primary | |
57 kDa |
Western Blot | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
CYPIIIA7, cytochrome P450, family 3, subfamily A, polypeptide 7, Cytochrome P450-HFLA, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, subfamily IIIA, polypeptide 7, xenobiotic monooxygenase | |
CYP3A7 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title