Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ D 4476
GREENER_CHOICE

Catalog No. p-4783891
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective CK1 inhibitor. Also inhibits TGF-βRI

Selective inhibitor of casein kinase 1 (CK1) and TGF-β type-I receptor (ALK5) that displays > 20-fold selectivity over SAPK2/p38 and a much greater selectivity over all other protein kinases tested. Suppresses site-specific phosphorylation and nuclear exclusion of FOXO1a.

Chemical Identifiers

CAS 301836-43-1
Molecular Formula C23H18N4O3

Specifications

Quantity 10 mg
Formula Weight 398.41
Purity >99%
Molecular Weight (g/mol) 398.422
Chemical Name or Material D 4476
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.