Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Altrose, 99%, Thermo Scientific™

Catalog No. AC258021000
Click to view available options
Quantity:
100 mg

Chemical Identifiers

CAS 1990-29-0
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00075620
InChI Key GZCGUPFRVQAUEE-UHFFFAOYNA-N
Synonym d-altro-hexose, 2s,3r,4r,5r-2,3,4,5,6-pentahydroxyhexanal, unii-5pln1o36ff, 5pln1o36ff, d-+-altrose, aldehydo-d-altrose
PubChem CID 94780
ChEBI CHEBI:28385
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal
SMILES OCC(O)C(O)C(O)C(O)C=O

Specifications

Melting Point 106°C to 108°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Merck Index 15, 317
Specific Rotation + 30
Solubility Information (10% in water) Clear colorless
Formula Weight 180.16
Percent Purity 99%
Physical Form Powder
Chemical Name or Material D(+)-Altrose, 99%
Show More Show Less
EINECSNumber 217-870-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.