Learn More
Tocris Bioscience™ D-AP5



Description
DL Mixture and L-isomer also available.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 100 mg |
| Formula Weight | 197.13 |
| Purity | >99% |
| Molecular Weight (g/mol) | 197.127 |
| Chemical Name or Material | D-AP5 |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.