Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ D-Serine
GREENER_CHOICE

Catalog No. 02261g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Potentiator of NMDA recepors; acts at glycine site

D-Serine is a glycine agonist at the NMDA receptor.

Chemical Identifiers

CAS 312-84-5
Molecular Formula C3H7NO3

Specifications

Quantity 1 g
Formula Weight 105.09
Purity >98%
Molecular Weight (g/mol) 105.093
Chemical Name or Material D-Serine
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.