Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ D609
GREENER_CHOICE

Catalog No. p-4783388
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 14-371-0

Supplier: Tocris Bioscience™ 1437/10

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective PC-PLC inhibitor

Selective competitive phosphatidyl choline-specific phospholipase C (PC-PLC) inhibitor (Ki = 6.4 μM); antiviral and antitumor agent. Also inhibits sphingomyelin synthase. Suppresses LPS- and IFNγ-induced NO production (IC50 = 20 mg/ml) and blocks oxidative glutamate toxicity in nerve cells. Antioxidant in vivo.

Chemical Identifiers

CAS 83373-60-8
Molecular Formula C11H18KOS2

Specifications

Quantity 10 mg
Formula Weight 266.46
Purity >98%
Molecular Weight (g/mol) 269.482
Chemical Name or Material D609
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.