Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DBX2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | DBX2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179220UL
![]() |
Novus Biologicals
NBP17921120UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179211
![]() |
Novus Biologicals
NBP179211 |
100 μL |
Each for $487.50
|
|
|||||
Description
DBX2 Polyclonal specifically detects DBX2 in Human samples. It is validated for Western Blot.Specifications
DBX2 | |
Polyclonal | |
Rabbit | |
NP_001004329 | |
440097 | |
Synthetic peptide directed towards the middle region of human DBX2The immunogen for this antibody is DBX2. Peptide sequence SSPRWRENSPEPSERLIQESSGAPPPEANSLQGALYLCSEEEAGSKGVLT. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
developing brain homeobox 2, developing brain homeobox protein 2, FLJ16139, homeobox protein DBX2 | |
DBX2 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title