Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCUN1D3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | DCUN1D3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15542320
![]() |
Novus Biologicals
NBP15542320UL |
20 μL |
Each for $206.00
|
|
|||||
NBP155423
![]() |
Novus Biologicals
NBP155423 |
100 μL |
Each for $487.50
|
|
|||||
Description
DCUN1D3 Polyclonal specifically detects DCUN1D3 in Human samples. It is validated for Western Blot.Specifications
DCUN1D3 | |
Polyclonal | |
Rabbit | |
Q8IWE4 | |
123879 | |
IgG | |
34 kDa |
Western Blot | |
Unconjugated | |
RUO | |
DCN1, defective in cullin neddylation 1, domain containing 3 (S. cerevisiae), DCN1-like protein 3, DCUN1 domain-containing protein 3, Defective in cullin neddylation protein 1-like protein 3, DKFZp686O0290, FLJ41725,44M2.4, MGC48972 | |
DCUN1D3 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title