Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decane, 99+%, pure

Catalog No. ac111871000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111871000 100 mL Glass bottle
AC111875000 500 mL Glass bottle
AC111870025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC111871000 Supplier Thermo Scientific Chemicals Supplier No. 111871000
Only null left
Add to Cart
This item is not returnable. View return policy

Decane, >-99.0%, C10H22, CAS Number-124-18-5, ccris 653, decane, analytical standard, decyl hydride, dsstox_cid_4913, dsstox_gsid_24913, dsstox_rid_77577, hsdb 63, n-decane, nonane, methyl, unii-nk85062oiy, 2.5L, 99+%, 01, 168, CHEBI:41808, Colorless, 0.7350g/mL, 204-686-4, 142.28, Pure, Danger | CAS: 124-18-5 | C10H22 | 142.286 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-18-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00008954
InChI Key DIOQZVSQGTUSAI-UHFFFAOYSA-N
Synonym n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
PubChem CID 15600
ChEBI CHEBI:41808
IUPAC Name decane
SMILES CCCCCCCCCC

Specifications

Color Colorless
Density (15°C) 0.733 to 0.737kg/L
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4103 to 1.4123 (20°C, 589nm)
Linear Formula CH3(CH2)8CH3
Quantity 100 mL
Beilstein 01, 168
Formula Weight 142.28
Percent Purity ≥99.0%
Grade Pure
Physical Form Liquid
Chemical Name or Material Decane, 99+%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-686-4
RTECSNumber HD6550000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.