Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dectin-1/CLEC7A Rabbit anti-Mouse, Clone: 10, Novus Biologicals™
SDP

Catalog No. p-7194775 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB024730 50 μg
NB024729 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB024730 Supplier Novus Biologicals Supplier No. NBP29049150UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Dectin-1/CLEC7A Monoclonal specifically detects Dectin-1/CLEC7A in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Dectin-1/CLEC7A
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 10
Conjugate Unconjugated
Dilution Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100
Formulation 0.2 um filtered solution in PBS
Gene Alias Beta-glucan receptor, BGR, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1
Host Species Rabbit
Immunogen Recombinant Mouse Dectin-1/CLEC7A protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64581
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.