Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dengue Virus 3 NS1 Mouse anti-Virus, Unconjugated, Clone: 411, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $457.74
Specifications
Antigen | Dengue Virus 3 NS1 |
---|---|
Clone | 411 |
Applications | ELISA, Flow Cytometry |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF21922MI
|
MilliporeSigma
MABF219225UL |
25 μL |
Each for $182.31
|
|
|||||
MABF21921MI
|
MilliporeSigma
MABF2192100UL |
100 μL |
Each for $457.74
|
|
|||||
Description
Dengue Virus 3 NS1 Monoclonal antibody specifically detects Dengue Virus 3 NS1 in Virus samples. It is validated for ELISA, Flow Cytometry.Specifications
Dengue Virus 3 NS1 | |
ELISA, Flow Cytometry | |
Unconjugated | |
Mouse | |
Virus | |
Purified recombinant Dengue virus 3 NS1 protein expressed in mammalian cells. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼48 kDa calculated for NS1 protein; 378.71 kDa for the entire genome polyprotein. |
411 | |
Monoclonal | |
Purified | |
Inflammation and Immunology | |
Denv3 NS1 protein | |
IgG1 κ | |
Protein G Purified | |
Clone 411 is a mouse monoclonal antibody that specifically detects D3engue virus 3 NS1 protein. It targets an epitope within 17 amino acids from the wing and finger regions. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title