Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Di-4-ANEPPS

Catalog No. d1199
Encompass_Preferred
Click to view available options
Quantity:
5 mg

Molecules that fluoresce in response to electrical potential changes in their environment

Increases and decreases in membrane potential-referred to as membrane hyperpolarization and depolarization, respectively-play a central role in many physiological processes, including nerve-impulse propagation, muscle contraction, cell signaling and ion-channel gating. Potentiometric probes are important tools for studying these processes.

  • These are fast-response probes that operate by means of change in their electronic structure, and consequently their fluorescence properties, in response to change in surrounding electric field
  • Their optical response is sufficiently fast to detect transient (millisecond) potential changes in excitable cells, including single neurons, cardiac cells and intact brains
  • However, magnitude of their potential-dependent fluorescence change is often small; fast-response probes typically show a 2-10% fluorescence change per 100mV
  • Furthermore, these dyes display potential-dependent shift in their excitation spectra, thus permitting quantitation of membrane potential using excitation ratio measurements
  • Zwitterionic molecule, exhibits most consistent potentiometric response in different cell and tissue types
  • Ex/Em maxima bound to model phospholipid membranes are approximately 465/635nm (but spectral properties are highly dependent on environment)
  • Nonfluorescent until bound to membranes
  • Soluble in ethanol, DMSO, and DMF
  • Dye is introduced into cells by direct addition of stock solution to cell culture medium or by using by retrograde labeling
  • Fast-response probe, suitable for detecting submillisecond membrane potential changes
  • Plasma membrane of cell typically has a transmembrane potential of approximately -70mV (negative inside) as a consequence of K+, Na+, and Cl- concentration gradients that are maintained by active transport processes
  • Potentiometric probes offer an indirect method of detecting the translocation of these ions

Cell Analysis, Cell Metabolism, Cell Structure, Cell Viability, Proliferation and Function, Membranes (General) and Lipids

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Content And Storage Store at room temperature and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type ANEP Dye
Quantity 5 mg
Shipping Condition Room Temperature
Sub Cellular Localization Cell Membranes & Lipids

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.