Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Dihydrexidine hydrochloride



Click to view available options
Quantity:
5 mg
25 mg
Description
A potent, full efficacy dopamine D1 agonist which shows no agonist activity at peripheral D2 receptors or adrenoceptors at doses which cause maximal stimulation of D1 sites. The compound appears to be fully bioavailable in brain and exhibits profound antiparkinsonism effects in vivo.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 5 mg |
| Formula Weight | 303.79 |
| Purity | >99% |
| Molecular Weight (g/mol) | 303.786 |
| Chemical Name or Material | Dihydrexidine hydrochloride |
Safety and Handling
| Recommended Storage | Desiccate at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction