Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Isolated from HeLa nuclear extracts as a complex consisting of a 400kDa catalytic subunit and a 155kDa heterodimeric DNA-binding component named Ku, which itself consists of subunits of approximately 85kDa and 70kDa
- Unit Definition: One unit is defined as the amount of enzyme required to transfer 1pmol of phosphate to DNA-Dependent Protein Kinase Peptide Substrate in 1 minute at 30°C
- Storage Buffer: 25mM HEPES (pH 7.5), 50mM KCl, 0.2mM EDTA, 10mM MgCl2, 1mM DTT, 10% glycerol
Specifications
Specifications
| Product Type | Protein Kinase |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction