Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNER Antibody, Novus Biologicals™
SDP

Catalog No. NBP17941520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17941520 20 μL
NBP179415 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17941520 Supplier Novus Biologicals Supplier No. NBP17941520UL

Rabbit Polyclonal Antibody

DNER Polyclonal specifically detects DNER in Human samples. It is validated for Western Blot.

Specifications

Antigen DNER
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. AAH24766
Gene Alias bet, delta and Notch-like epidermal growth factor-related receptor, delta/notch-like EGF repeat containing, delta-notch-like EGF repeat-containing transmembrane, H_NH0150O02.1, UNQ26, WUGSC:H_NH0150O02.1
Gene Symbols DNER
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human DNERThe immunogen for this antibody is DNER. Peptide sequence DACQRKPCQNNASCIDANEKQDGSNFTCVCLPGYTGELCQSKIDYCILDP.
Purification Method Protein A purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 92737
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.