Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOT1L Rabbit anti-Human, Mouse, Clone: 7A10H9, Novus Biologicals™
SDP

Catalog No. p-7246972 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165989 20 μg
NB165988 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB165989 Supplier Novus Biologicals Supplier No. NBP31544620UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DOT1L Monoclonal antibody specifically detects DOT1L in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen DOT1L
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 7A10H9
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias DOT1-like protein, DOT1-like, histone H3 methyltransferase (S. cerevisiae), EC 2.1.1.43, H3-K79-HMTase, KIAA1814DKFZp586P1823, KMT4H3 lysine-79 specific, Lysine N-methyltransferase 4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human DOT1L (Q8TEK3).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Chromatin Research, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 84444
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.