Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPYD Antibody (SR1055), Novus Biologicals™
SDP

Catalog No. NB258295 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258295 100 μg
NB258059 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258295 Supplier Novus Biologicals Supplier No. NBP322329100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

DPYD Monoclonal antibody specifically detects DPYD in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen DPYD
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone SR1055
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias DHP, DHPDHase, dihydropyrimidine dehydrogenase, dihydropyrimidine dehydrogenase [NADP+], Dihydrothymine dehydrogenase, Dihydrouracil dehydrogenase, DPDMGC132008, EC 1.3.1.2, MGC70799
Host Species Rabbit
Immunogen A synthesized peptide derived from human DPYD (Uniprot #: Q12882)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1806
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.