Learn More
Dräger™ Pac™ 5500 Single-Gas Detector
Unlimited instrument life
Specifications
Alarm Types | Audible, visual, vibrating |
---|---|
Battery Life | 8 hr. |
Depth (English) | 1 in. |
Depth (Metric) | 2.5 cm |
Height (English) | 2.5 in. |
Includes: Alligator clip for secure attachment
Catalog Number | Mfr. No. | Components Measured | Includes | Measurement Range | Response Time | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Components Measured | Includes | Measurement Range | Response Time | Price | Quantity & Availability | ||
17-985-680
|
Dräger™
4543960 |
Carbon Monoxide | Alligator clip | 0 to 500 | 15 sec. | N/A | N/A | N/A | |
17-985-684
|
Dräger™
8322013 |
Oxygen | Blue keypad overlay, Alligator clip | 0 to 25% | 10 sec. | N/A | N/A | N/A | |
Description
These hardy sensors can be used in extreme environmental conditions without triggering false alarms.
- Compact, robust and ergonomic design with intuitive two-button operation
- Ideal for personal monitoring of industrial oxygen, carbon monoxide, or hydrogen sulfide
- Large, language-free backlit display continuously indicates gas concentration while simultaneously displaying the corresponding measurement unit
- Equipped with a sensor that allows gas intake from both the top and front of the unit to ensure continuous measurement, even if a gas inlet is covered
- Triple alarms for exceptional safety—clear, multi-toned signal, a bright, flashing 360° LED on top and bottom, and vibration
- Store up to 60 events, along with dates and times; infrared interface enables data to be loaded onto a PC for analysis (requires connecting cradle or E-Cal System and software, sold separately)
- When bump mode is activated, the unit automatically reminds users when a bump test or calibration is required
- Operates in a temperature range from -22° to +122°F (-30° to +50°C); relative humidity, 10 to 90%
Order Info
Dimensions: 0.8L x 2.5W x 3.3 in. H (2.0 x 6.4 x 8.4cm). Weight: 3.8 oz. (106g).
Certifications
IP65 rated for ingresss protection. UL Classified as Intrinsically Safe for use in Class I, Division 1, Group A, B, C, D, E, F, G hazardous locations, Temp Code T4; ATEX, I/II 1/2 G EEx ia I/IIC, T4. CE mark.
Compliance
Complies with EMC Directive 89/336/EEC.
Specifications
Audible, visual, vibrating | |
1 in. | |
2.5 in. | |
10 to 90% | |
700 to 1300hpa | |
-30°C to +50°C | |
106g | |
8.4 cm |
8 hr. | |
2.5 cm | |
6.4 cm | |
65 | |
-20°F to +120°F | |
3.8 oz. | |
3.3 in. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.