Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUOX2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11061576 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11061576 0.1 mL
NB11061576T 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11061576 Supplier Novus Biologicals Supplier No. NB11061576
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

DUOX2 Polyclonal specifically detects DUOX2 in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen DUOX2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin reported in scientific literature (PMID 24492313), KnockDown Validated
Formulation PBS and 30% Glycerol with 0.1% Sodium Azide
Gene Accession No. Q9NRD8
Gene Alias dual oxidase 2, dual oxidase-like domains 2, EC 1.11.1.-, EC 1.6.3.1, flavoprotein NADPH oxidase, Large NOX 2, LNOX2P138(TOX), Long NOX 2, NADH/NADPH thyroid oxidase p138-tox, NADPH oxidase/peroxidase DUOX2, NADPH thyroid oxidase 2, nicotinamide adenine dinucleotide phosphate oxidase, NOXEF2, p138 thyroid oxidase, P138-TOX, THOX2TDH6, Thyroid oxidase 2
Gene Symbols DUOX2
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human DUOX2 (within residues 400-500). [Swiss-Prot# Q9NRD8]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Lipid and Metabolism, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 50506
Target Species Human, Mouse, Rat, Pig, Canine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.