Learn More
Thermo Scientific™ EASY-Spray™ PepMap™ Neo UHPLC Columns
Description
Separate challenging peptide mapping samples using Thermo Scientific™ EASY-Spray™ PepMap™ Neo UHPLC columns. These columns feature easy connectivity, high reproducibilty, and excellent separations. Our Neo columns provide 1500 bar pressure capability, improved column-to-column consistency, and increased efficiency. The column media is manufactured and selected to exacting standards and packed at high pressure, resulting in enhanced peak symmetry, resolution, and column-to-column reproducibility that allows you to obtain greater sample coverage and sample insights. The Click-and-Spray format of EASY-Spray columns enables perfect connections to our mass spectrometers every time.
Benefits of PepMap Neo Columns
- Brilliant Connectivity — EASY-Spray™ columns click into position with ease.
- Brilliant Reproducibility — Precise manufacturing protocols for packing media improve column-to-column consistency for reproducible analysis every day.
- Brilliant Separation — Ultra-high pressure packing increases column efficiency, allowing you to run at up to 1500 bar for enhanced peak symmetry and resolution, providing higher sample capacity for more sample insights.
Specifications
Specifications
| Column Format | Analytical Column, EASY-Spray™ |
| Diameter (Metric) | 75 μm |
| Length (Metric) | 150 mm |
| Max. Pressure | 22,000 psi (1500 bar) |
| Particle Size | 2 μm |
| pH | 2 to 8 |
| Pore Size | 100 Å |
| Surface Area | 450 m2/g |
| Temperature | 60°C |
| USP Type | L1 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.