Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECHDC3 Antibody, Novus Biologicals™
SDP

Catalog No. p-7109202 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP179302 100 μL
NBP17930220 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP179302 Supplier Novus Biologicals Supplier No. NBP179302
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ECHDC3 Polyclonal specifically detects ECHDC3 in Human samples. It is validated for Western Blot.

Specifications

Antigen ECHDC3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. EAW86340
Gene Alias enoyl CoA hydratase domain containing 3, enoyl Coenzyme A hydratase domain containing 3, FLJ20909, mitochondrial
Gene Symbols ECHDC3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal of human ECHDC3The immunogen for this antibody is ECHDC3. Peptide sequence SLAMLKSLQSDILHDADSNDLKVIIISAEGPVFSSGHDLKELTEEQGRDY.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79746
Test Specificity Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Chicken: 92%; Xenopus: 85%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit, Yeast, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.