Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGFR Antibody (Matuzumab) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. N27591102M Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N27591102M 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N27591102M Supplier Novus Biologicals Supplier No. NBP2759110.2MG
Only null left
Add to Cart
Add to Cart

Human Monoclonal Antibody

EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Block/Neutralize.

Specifications

Antigen EGFR
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Blocking Assay, Neutralization
Classification Monoclonal
Clone Matuzumab
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Block/Neutralize
Formulation PBS with 0.02% Proclin 300
Gene Alias avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
Gene Symbols EGFR
Host Species Human
Immunogen Domain III of the extracellular domain of human EGRF.
Quantity 0.2 mg
Primary or Secondary Primary
Gene ID (Entrez) 1956
Test Specificity Binds to human EGFR.
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.