Learn More
Description
Specifications
Specifications
| Antigen | EGLN3/PHD3 |
| Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Simple Western 1:1000, Chromatin Immunoprecipitation reported in scientific literature (PMID 21620138), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 20801873), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 15721254), Immunohistochemistry-Paraffin reported in scientific literature (PMID 25161887), Immunoblotting, Chromatin Immunoprecipitation (ChIP) |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | Q9H6Z9 |
| Gene Alias | EC 1.14.11, EC 1.14.11.-, EGL nine (C.elegans) homolog 3, egl nine homolog 3, egl nine homolog 3 (C. elegans), egl nine-like protein 3 isoform, FLJ21620, HIF prolyl hydroxylase 3, HIF-PH3, HIFPH3 MGC125998, HIF-prolyl hydroxylase 3, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 3, MGC125999, pdh3, PHD3, Prolyl hydroxylase domain-containing protein 3 |
| Gene Symbols | EGLN3 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
