Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF3G Antibody, Novus Biologicals™
SDP

Catalog No. p-5067430 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224524SS 0.025 mg
NBP224524 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224524SS Supplier Novus Biologicals Supplier No. NBP224524SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EIF3G Polyclonal specifically detects EIF3G in Human, Mouse, Rat, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EIF3G
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.025-0.1 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. O75821
Gene Alias eIF3 p42, eIF3 p44, eIF-3-delta, eIF3-delta, eIF3g, EIF3-P42, eIF3-p44, EIF3S4eIF-3 RNA-binding subunit, Eukaryotic translation initiation factor 3 RNA-binding subunit, Eukaryotic translation initiation factor 3 subunit 4, eukaryotic translation initiation factor 3 subunit G, eukaryotic translation initiation factor 3 subunit p42, eukaryotic translation initiation factor 3, subunit 4 (delta, 44kD), eukaryotic translation initiation factor 3, subunit 4 delta, 44kDa, eukaryotic translation initiation factor 3, subunit G
Gene Symbols EIF3G
Host Species Rabbit
Immunogen A portion of amino acids 200-250 of human eIF3G was used as the immunogen for the antibody.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8666
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in human, monkey and dog, 92% homologous in horse and chimpanzee, and 76% homologous in cow.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.