Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Force Sensor
SCI_ED

Catalog No. S13459 Shop All Eisco Products
Click to view available options
Type:
Sensor, Force

Requires

Eisco Sensors Base Unit

Demonstrates force, gravity, and energy.

  • Enables both pushing and pulling experiments so students can see, feel, and understand force concepts while measuring data digitally
Specifications:
  • Range: -10N to 10N; -50N to 50N
  • Resolution: 0.01N
  • Sample Rate: 3000 S/sec
Applications:
  • Beginning force measurements, Spring forces, Friction analysis, Gravitational force analysis

Specifications

Type Sensor, Force
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.