Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Requires
Eisco Sensors Base Unit
- Two operational modes make it easy to measure both sound-pressure (decibels) and frequency
- Range: 40 to 110dB, 0 to 4,095 Signal
- Resolution: 0.1dB, 1
- Sample Rate: 10,000 S/sec
- Measuring noise level of classroom, examining frequency of car horn, studying nature of sound waves
Specifications
Specifications
| Type | Sensor, Sound |
| For Use With (Application) | Measuring noise level of classroom, examining frequency of car horn, studying nature of sound waves |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
